Learn More
WDR92 Monoclonal Antibody (OTI2F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809522S
Encompass_Preferred
 View more versions of this product
Product Code. 17591844
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
WDR92 Monoclonal antibody specifically detects WDR92 in Human samples. It is validated for Western Blot
Specifications
Specifications
WDR92
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96MX6
Mouse
Affinity chromatography
RUO
116143
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Western Blot
OTI2F2
Unconjugated
WDR92
AI553587; HZGJ; monad; RGD1563863; testicular secretory protein Li 67; WD repeat domain 92; WD repeat-containing protein 92; WD repeat-containing protein Monad; Wdr92
Full length human recombinant protein of human WDR92 produced in HEK293T cell.
30 μL