missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR92 Monoclonal Antibody (OTI4B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809523
This item is not returnable.
View return policy
Description
WD40 repeats are a common structural module in eukaryotic proteins, and proteins containing WD40 domains have a wide range of functions, including signal transduction, cell cycle regulation, RNA splicing, and transcription. One such protein, WDR92, also known as monad, contains two WD40 repeats is widely expressed in human tissues, especially testis. Overexpression of this protein or its binding partner RNA polymerase II-associated protein 3 (RPAP3) potentiated apoptosis and caspase-3 activation induced by TNF-a and cycloheximide, suggesting that WDR92, together with RPAP3, may function as a novel modulator of apoptosis.Specifications
| WDR92 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96MX6 | |
| WDR92 | |
| Full length human recombit protein of human WDR92 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI4B9 | |
| Unconjugated | |
| WDR92 | |
| WD repeat domain 92 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 116143 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction