missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WEHI-345 analog, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Description
WEHI-345 analog is the analog of WEHI-345. WEHI-345 is a potent and selective RIPK2 kinase inhibitor with an IC50 of 0.13 μM, which delays RIPK2 ubiquitylation and NF-κB activation on oligomerization domain (NOD) stimulation.
Specifications
Specifications
| Chemical Name or Material | WEHI-345 analog |
| CAS | 1354825-62-9 |
| Color | Gray |
| Purity Grade Notes | Research |
| Molecular Formula | C23H25N7O |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 30 mg/mL (72.20 mM) |
| SMILES | O=C(C1=NC=CC=C1C)NCC(C)(N2N=C(C3=CC=C(C)C=C3)C4=C(N)N=CN=C42)C |
| Molecular Weight (g/mol) | 415.49 |
| Formula Weight | 415.49 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction