missing translation for 'onlineSavingsMsg'
Learn More

WEHI-345 analog, MedChemExpress

Product Code. 16419476
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16419476

Brand: Medchem Express HY100112

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WEHI-345 analog is the analog of WEHI-345. WEHI-345 is a potent and selective RIPK2 kinase inhibitor with an IC50 of 0.13 μM, which delays RIPK2 ubiquitylation and NF-κB activation on oligomerization domain (NOD) stimulation.

WEHI-345 analog is the analog of WEHI-345. WEHI-345 is a potent and selective RIPK2 kinase inhibitor with an IC50 of 0.13 μM, which delays RIPK2 ubiquitylation and NF-κB activation on oligomerization domain (NOD) stimulation.

Specifications

Chemical Name or Material WEHI-345 analog
CAS 1354825-62-9
Color Gray
Purity Grade Notes Research
Molecular Formula C23H25N7O
Quantity 50 mg
Solubility Information DMSO : ≥ 30 mg/mL (72.20 mM)
SMILES O=C(C1=NC=CC=C1C)NCC(C)(N2N=C(C3=CC=C(C)C=C3)C4=C(N)N=CN=C42)C
Molecular Weight (g/mol) 415.49
Formula Weight 415.49
Percent Purity 99.47%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.