Learn More
Grant Instruments Well Plate Adaptor
Product Code. 30274371
Click to view available options
Type:
348-well PCR Plate Adapter
96-well PCR Plate Adapter
Unit Size:
Pack of 2
Product Code. 30274371
Brand: Grant Instruments AP384
Centrifuge rotor adapter for well plates.
  • Set of 2 adaptors
  • To be used with R-2 and Ri-2 rotors
  • Rotors can be used with LMC-56, LMC-3000 and LMC-4200R
  • Adaptors available for 96-well plates or 384-well plates
TRUSTED_SUSTAINABILITY
Specifications
Type 348-well PCR Plate Adapter
Product Line Grant-Bio LMC Centrifuges
Material Plastic
Capacity 2 x 384-well Plates
Autoclavable Autoclavable
For Use With (Equipment) Grant Instruments Centrifuges
Color White
Max. RCF 1,250 x g
Max. Speed 2,000 rpm
Length (Metric) 124 mm
Height (Metric) 14 mm
Width (Metric) 86 mm
No. per Pack 2/Pk.