missing translation for 'onlineSavingsMsg'
Learn More
Learn More
West Nile Virus C Protein Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 610.00
Specifications
| Antigen | West Nile Virus C Protein |
|---|---|
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Form | Purified |
Description
West Nile Virus C Protein Polyclonal antibody specifically detects West Nile Virus C Protein in Virus samples. It is validated for ELISASpecifications
| West Nile Virus C Protein | |
| Polyclonal | |
| Purified | |
| RUO | |
| Virus | |
| Capside protein C, Core protein, West Nile Core, West Nile Virus Core protein, WNV core | |
| Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the amino-terminus of the WNV Core protein. The immunogen is located within the first 50 amino acids of WNV Core . Amino Acid Squence: SKKPGGPGKNRAVNM | |
| Primary | |
| Store at 4°C in the dark. |
| ELISA | |
| mFluor Violet 610 SE | |
| Rabbit | |
| Immunology, Virology Bacteria and Parasites | |
| 50mM Sodium Borate | |
| 912267 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title