missing translation for 'onlineSavingsMsg'
Learn More
Learn More
West Nile Virus Envelope Antibody (15), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06463UV
This item is not returnable.
View return policy
Description
West Nile Virus Envelope Monoclonal antibody specifically detects West Nile Virus Envelope in Virus samples. It is validated for Western Blot
Specifications
| West Nile Virus Envelope | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant West Nile Virus (lineage 1, strain NY99) Envelope Protein (Domain III) (ABA62343.1; Gln586-Ala705). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| 15 | |
| Western Blot | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction