missing translation for 'onlineSavingsMsg'
Learn More
Learn More
West Nile Virus MP Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41053V
This item is not returnable.
View return policy
Description
West Nile Virus MP Polyclonal antibody specifically detects West Nile Virus MP in Virus samples. It is validated for ELISA
Specifications
| West Nile Virus MP | |
| Polyclonal | |
| ELISA | |
| developmental pluripotency associated 3, DPPA3, DPPA3 developmental pluripotency associated 3 | |
| Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the carboxy terminus of the West Nile Virus MP. The immunogen is located within amino acids 220 - 270. Amino Acid Squence: LDSTKATRYLVKTES | |
| 0.1 mL | |
| Immunology, Virology Bacteria and Parasites | |
| Virus, Mouse, Rat | |
| Purified |
| ELISA | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction