missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ West Nile Virus NS1 Protein Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111988
This item is not returnable.
View return policy
Description
West Nile Virus NS1 Protein Polyclonal Antibody for Western Blot
West Nile Virus (WNV) is a member of the Flaviviridae, a plus-stranded virus family that includes St. Louis encephalitis virus, yellow fever virus, and Dengue virus. WNV was initially isolated in 1937 in the West Nile region of Uganda and has become prevalent in Africa, Asia, and Europe. It has rapidly spread across the United States with cases being observed in every continental state. Virus particles consist of a dense core made up of the core/capsid protein encapsulating the RNA genome surrounded by a membrane envelope embedded with envelope and matrix proteins which play a major role for WNV entry into target cells. The viral core protein is thought to contribute to the WNV-associated inflammation via apoptosis induced though the caspase-9 pathway as delivery of core gene delivery into the striatum of mouse brain and skeletal muscle resulted in cell death and inflammation.
Specifications
| West Nile Virus NS1 Protein | |
| Polyclonal | |
| Unconjugated | |
| West Nile Virus | |
| Recombinant protein encompassing a sequence within the center region of West Nile virus NS1 protein (West Nile virus (strain NY99-IC)). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 2.57 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction