missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WFDC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116731
This item is not returnable.
View return policy
Description
Positive Samples: 293T, SGC-7901, Mouse brain, Mouse lung Immunogen sequence: PPVLDWLVQP KPRWLGGNGW LLDGPEEVLQ AEACSTTEDG AEPLLCPSGY ECHILSPGDV AEGIPNRGQC VKQRRQADGR ILRHKLYKEY PEGDSKNVAE PGRGQQKHFQ.
This gene encodes a member of the WAP-type four disulfide core domain family. The WAP-type four-disulfide core domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. The encoded protein shares 81% amino acid identity with the rat ps20 protein, which was originally identified as a secreted growth inhibitor. This gene is mapped to chromosome 16q24, an area of frequent loss of heterozygosity in cancers, including prostate, breast and hepatocellular cancers and Wilms' tumor. Owing to its location and a possible growth inhibitory property of its gene product, this gene is suggested to be a tumor suppressor gene.
Specifications
| WFDC1 | |
| Polyclonal | |
| Unconjugated | |
| WFDC1 | |
| 2310058A03Rik; Prostate stromal protein ps20; PS20; ps20 growth inhibitor; WAP four-disulfide core domain 1; WAP four-disulfide core domain 1 homolog; WAP four-disulfide core domain protein 1; Wfdc1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 58189, 67866 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.42 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9ESH5, Q9HC57 | |
| WFDC1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 111-220 of human WFDC1 (NP_0670202). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction