missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WFDC2 Monoclonal Antibody (OTI3A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801084S
This item is not returnable.
View return policy
Description
WFDC2 Monoclonal antibody specifically detects WFDC2 in Human samples. It is validated for Western BlotSpecifications
| WFDC2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P19971 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1890 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3A9 | |
| Unconjugated | |
| TYMP | |
| 1600023A02Rik; dJ461P17.6; EDDM4; epididymal protein 4; epididymal secretory protein 4; Epididymal secretory protein E4; epididymis-specific, whey-acidic protein type, four-disulfide core; HE4; Major epididymis-specific protein E4; MGC57529; putative protease inhibitor WAP5; RE4; WAP domain containing protein HE4-V4; WAP domain-containing protein HE4; WAP four-disulfide core domain 2; WAP four-disulfide core domain protein 2; WAP5; Wfdc2 | |
| Human recombinant protein fragment corresponding to amino acids 311-482 of human TYMP produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction