missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WFDC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551168
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total WFDC5 protein.
WFDC5 is a putative acid-stable proteinase inhibitor.This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. Most WFDC proteins contain only one WFDC domain, and this encoded protein contains two WFDC domains. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. Most WFDC gene members are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the centromeric cluster.
Specifications
| WFDC5 | |
| Polyclonal | |
| Unconjugated | |
| WFDC5 | |
| BC019734; dJ211D12.5; p53-responsive gene 5 protein; Prg5; protease inhibitor WAP1; putative protease inhibitor WAP1; UNQ3036/PRO9842; WAP four-disulfide core domain 5; WAP four-disulfide core domain protein 5; WAP1; WFDC5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 149708, 209232 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 3.5 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8TCV5 | |
| WFDC5 | |
| Fusion protein of human WFDC5. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction