missing translation for 'onlineSavingsMsg'
Learn More

WFDC8 Antibody, Novus Biologicals™

Product Code. 18678087 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μL
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18678087 25 μL 25µL
18608848 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18678087 Supplier Novus Biologicals Supplier No. NBP26260825ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WFDC8 Polyclonal antibody specifically detects WFDC8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen WFDC8
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias C20orf170, dJ461P17.1, WAP8, WFDC8 WAP four-disulfide core domain 8
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: KHKPGLCPKERLTCTTELPDSCNTDFDCKEYQKCCFFACQKKCMDPFQEPCMLPVRHGNCNHEAQRWHFDFKNYRCTPFKYRGCEGNA
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90199
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.