missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WFDC8 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA584857

Product Code. 16361805

  • € 524.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GQCPLFPFTE RKECPPSCHS DIDCPQTDKC CESRCGFVCA RAWTVKKGFC PRKPLLCTKI DKPKCLQDEE CPLVEKCCSH CGLKCMDPR Highest antigen sequence indentity to the following orthologs: Mouse - 73%, Rat - 73%.

This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. The encoded protein contains a Kunitz-inhibitor domain, in addition to three WFDC domains. Most WFDC genes are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the telomeric cluster. Two alternatively spliced transcript variants have been found for this gene, and they encode the same protein. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

WFDC8
Polyclonal
Unconjugated
WFDC8
C20orf170; dJ461P17.1; epididymis secretory protein Li 292; HEL-S-292; protease inhibitor WAP8; putative protease inhibitor WAP8; testicular secretory protein Li 68; WAP four-disulfide core domain 8; WAP four-disulfide core domain protein 8; WAP motif protein 1; WAP8; WFDC8
Rabbit
Antigen affinity chromatography
RUO
90199
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IUA0
WFDC8
Recombinant protein corresponding to Human WFDC8. Recombinant protein control fragment (Product #RP-108252).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.