missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WFS1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94845-0.1ml
This item is not returnable.
View return policy
Description
WFS1 Polyclonal antibody specifically detects WFS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| WFS1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| DFNA14, DFNA38, DFNA6, DIDMOAD, FLJ51211, WFRS, WFSwolframin, Wolfram syndrome 1 (wolframin) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-285 of human WFS1 (NP_005996.2). MDSNTAPLGPSCPQPPPAPQPQARSRLNATASLEQERSERPRAPGPQAGPGPGVRDAAAPAEPQAQHTRSRERADGTGPTKGDMEIPFEEVLERAKAGDPKAQTEVGKHYLQLAGDTDEELNSCTAVDWLVLAAKQGRREAVKLLRRCLADRRGITSENEREVRQLSSETDLERAVRKAALVMYWKLNPKKKKQVAVAELLENVGQVNEHDGGAQPGPVPKSLQKQRRMLERLVSSESKNYIALDDFVEITKKYAKGVIPSSLFLQDDEDDDELAGKSPEDLPLR | |
| 0.1 mL | |
| Cellular Markers, ER Markers, Lipid and Metabolism, Neuroscience, Sensory Systems, Signal Transduction, Vision | |
| 7466 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction