missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WHAMM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558886
This item is not returnable.
View return policy
Description
Immunogen sequence: RALSSSSQAA THQNLGFRAP VKDDQPRPLV CESPAERPRD SLESFSCPGS MDEVLASLRH GRAPLRKVEV PAVRPPHASI NEHILAAIR Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 54%.
Specifications
| WHAMM | |
| Polyclonal | |
| Unconjugated | |
| WHAMM | |
| KIAA1971; WAS protein homolog associated with actin, golgi membranes and microtubules; WAS protein homology region 2 domain-containing protein 1; WASP homolog-associated protein with actin, membranes and microtubules; WH2 domain-containing protein 1; WHAMM; WHAMM1; WHDC1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 123720 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TF30 | |
| WHAMM | |
| Recombinant protein corresponding to Human WHAMM. Recombinant protein control fragment (Product #RP-98104). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction