Learn More
Cytiva Whatman Cyclopore Polycarbonate Membrane
Click to view available options
Pore Size:
0.2 μm
0.4 μm
0.5 μm
5 μm
Unit Size:
Pack of 100
Pack of 25
Dimensions (L x W):
209.55 x 292.1 mm
25 x 80 mm
Diameter (Metric):
293 mm
Description
- Low affinity for stains improves visibility under microscope
- Low protein binding minimizes sample loss of proteinaceous samples
- Narrow pore size distribution and high pore density support rapid surface capture of analyte
- Negligible absorption and adsorption of filtrate; nonhygroscopic
- No particle shedding enables clean filtrate
- Biologically inert
Specifications
Specifications
| Material | Polycarbonate |
| Pore Size | 5 μm |
| Quantity | 100 Pk. |