Learn More
QIAGEN Multi-Barrier Pouches
For contamination-free collection, storage and transport of DNA samples.
Brand: QIAGEN WB100037
Additional Details : Weight : 0.58000kg
Description
For contamination-free collection, storage and transport of DNA samples.
- Multi-barrier Pouches for transport or storage of QIAcard FTA Mini and Micro formats.
- Laminated layers maintain sample integrity and security by protecting the card from exposure to gas or liquid contamination.
- Maintain sample security by tamper-evident seal.
- The outer paper surface is suitable for labeling or writing.
- To optimize sample integrity FTA Cards are stored in a Multi-barrier Pouch with a desiccant packet.
Specifications
FTA classic card | |
Multi-barrier Pouch | |
Medium (11.1 x 16.5 cm) |
Collection, storage, processing and shipping FTA™ and FTA™ elute cards | |
100 Pk. |