Learn More
QIAGEN QIAcard FTA Mini
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Mini with 2 sample areas per card.
Brand:  QIAGEN WB120355
Additional Details : Weight : 0.06000kg
 View more versions of this product
Product Code. 11969075
  • € 149.00 / Pack of 25
Estimated Shipment: 15-04-2024
to see stock.
Description
Description
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Mini with 2 sample areas per card.
  • QIAcard FTA Mini format with 2 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
  • FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Specifications
Specifications
Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Mini Cards.
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
FTA Mini Card
125 μL
Blood
2
25 Pk.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning