Learn More
QIAGEN QIAcard FTA Indicating Mini
Click to view available options
Quantity:
100 Pk.
25 Pk.
Unit Size:
Pack of 100
Pack of 25
Description
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Mini with 2 sample areas per card.
- QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
- Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
- Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Specifications
| Description | Collect, stabilize, process, transport, and archive buccal DNA samples with Indicating FTA Micro Cards. |
| Sample Type | Buccal Cells, Saliva |
| For Use With (Application) | Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification,Molecular biology |
| No. of Samples | 2 |
| Product Type | Indicating FTA Mini Card |
| Quantity | 100 Pk. |
| Sample Volume | 125 μL |
Safety and Handling
Signal Word
- Warning