Learn More
Cytiva Whatman™ Grade 470
Product Code. 11338944
Click to view available options
Dimensions (L x W):
203 x 305 mm
450 ft x 1.5 in.
460 x 570 mm
Unit Size:
Each
Pack of 100
Pack of 25
Product Code. 11338944
Brand: Cytiva 10318493
Only null left
Add to basket
Add to basket
For electrophoresis and amino acid chromatography
  • Soft surface
  • For gelatinous samples
  • For the absorption of culture media
  • Used as blotting paper and amino acid chromatography
TRUSTED_SUSTAINABILITY
Specifications
Format Sheet
Grade 470
Quantity 100/Pk.
Dimensions (L x W) 460 x 570 mm
Length (Metric) 460 mm
Width (Metric) 570 mm