Learn More
Cytiva Whatman™ Nuclepore Track-Etched Polycarbonate AOX Membranes
Click to view available options
Diameter (Metric):
25 mm
47 mm
Unit Size:
Pack of 100
Description
- High chemical resistance and thermal stability for a wide range of samples
- Low, consistent ash levels and low tare weights
- Compatible with ISO 9562 guidelines for determining adsorbable organically bound halogens
- Smooth flat surface increases particle visibility
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Nucleopore Polycarbonate |
| Pore Size | 0.4 μm |
| Flow Rate | High |
| For Use With (Application) | Epifluorescence microscopy, environmental analysis, cell biology |
| Hydrophobicity | Hydrophilic |
| Diameter (Metric) | 25 mm |