Learn More
DWK Life Sciences Wheaton™ Amber Boston Rounds with PE Cone Lined Caps
Product Code. 12369769 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Closure Size:
20-400 mm
22-400 mm
24-400 mm
28-400 mm
33-400 mm
Unit Size:
Pack of 12
Pack of 24
Pack of 48
Product Code. 12369769
Brand: DWK Life Sciences W216853
Only null left
Add to basket
Add to basket
Amber Type III soda lime glass protects light-sensitive solutions
  • Narrow mouth enables liquids to be carefully poured to prevent spillage
TRUSTED_SUSTAINABILITY
Specifications
Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Amber
Diameter (English) 3.7 in.
Material Soda Lime Glass
Closure Color Black
Height (Metric) 208 mm
Height (English) 8.18 in.
Closure Liner Polyethylene
Closure Material Phenolic
Closure Size 33-400 mm
Diameter (Metric) 94 mm
Closure Type Screw Cap