Learn More
DWK Life Sciences Wheaton™ Clear Boston Rounds with TFE-lined Caps
Product Code. 10661233 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Material:
Glass
Soda Lime Glass
Unit Size:
Pack of 12
Pack of 24
Pack of 48
Product Code. 10661233
Brand: DWK Life Sciences W216807
Only null left
Add to basket
Add to basket
Ideal for solvent, chemical and sample storage
  • Narrow mouth enables liquids to be carefully poured to prevent spillage
Order Info
Available with caps attached or without caps (order caps separately).
Compliance
USP Type-III soda-lime glass
TRUSTED_SUSTAINABILITY
Specifications
Capacity 2 oz., 60 mL
Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Clear
Diameter (English) 1.53 in.
Material Soda Lime Glass
Closure Color White
Certifications/Compliance EPA, FDA
Height (Metric) 94 mm
Height (English) 3.7 in.
Closure Liner Polyethylene
Closure Material Polypropylene
Closure Size 20-400 mm
Diameter (Metric) 39 mm
Closure Type Screw Cap