Learn More
DWK Life Sciences Wheaton™ Clear French Squares with PTFE Faced PE-lined Closures
Product Code. 10633523 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
15 mL
30 mL
60 mL
125 mL
250 mL
500 mL
Unit Size:
Pack of 24
Pack of 48
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10633523 125 mL Pack of 24
10396892 15 mL Pack of 48
10501043 30 mL Pack of 48
10483322 60 mL Pack of 48
10255112 250 mL Pack of 24
10693683 500 mL Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10633523 Supplier DWK Life Sciences Supplier No. W216880
Only null left
Add to basket
Edge
Add to basket
Square shape maximizes lab storage space
  • USP Type III soda-lime glass
  • Ideal for solvent, chemical or sample storage
TRUSTED_SUSTAINABILITY
Specifications
Capacity 4 oz., 125 mL
Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Clear
Diameter (English) 1.77 in.
Material Soda Lime Glass
Closure Color White
Height (Metric) 113 mm
Height (English) 4.44 in.
Closure Liner PTFE/Polyethylene
Closure Material Polypropylene
Closure Size 33-400 mm
Diameter (Metric) 45 mm
Closure Type Screw Cap