Learn More
DWK Life Sciences Wheaton™ Clear Straight-Sided Jars without Caps
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
Closure Size:
53-400 mm
58-400 mm
70-400 mm
89-400 mm
Unit Size:
Pack of 12
Pack of 144
Pack of 24
Description
Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass
- Wide mouths provide easy access to contents when using spoons or spatulas
- Chemical-resistant caps conform to EPA and FDA regulations
- Available in partitioned Convenience or Valu-Bulk™ cartons
- Each case of Convenience Pack Bottles includes Hazard Communication labels for proper content identification and Tamper
- Evident labels to protect sample integrity in transit or long-term storage
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Capacity (English) | 16 oz. |
| Color | Clear |
| Material | Soda Lime Glass |
| Height (Metric) | 95 mm |
| Height (English) | 3.7 in. |
| Closure Size | 89-400 mm |
| Diameter (Metric) | 91 mm |
| Closure Type | Screw Cap |