Learn More
DWK Life Sciences Wheaton™ Glass Staining Dishes for 50 Slides
Shop All Wheaton Science Products Products
Click to view available options
Includes:
Cover
Dish
Dish, Cover
Dish, Cover, Rack
Rack
Unit Size:
Pack of 3
Pack of 6
Description
- Dish and cover are made from soda-lime glass
- Dimensions: 3.25 L × 7.875 W × 4.125 in. H (80 × 200 × 105 mm)
- Nontarnishable and withstands most staining solutions
- Holds up to 50 slides horizontally
- Hinged handle allows easy insertion and removal of slides
Specifications
Specifications
| Closure Included | Yes |
| Material | Soda Lime Glass |
| Type | Cover |
| For Use With (Equipment) | Glass staining dish for 50 slides |
| Includes | Cover |