Learn More
DWK Life Sciences Wheaton™ Hose Connector
Shop All Wheaton Science Products Products
Click to view available options
Thread Size:
13-425 GPI
Unit Size:
Pack of 6
Description
- For continuous extraction of analytes
- All wetted parts are TFE
- Unscrew to disconnect, leaving hose attached
- O.D of smallest serration:0.375 in. (9.5mm)
Specifications
Specifications
| Certifications/Compliance | USP Type I, ASTM E438, Type I, Class A |
| Material | PTFE |
| For Use With (Application) | For continuous extraction of analytes and micro soxhlet extraction apparatus |
| Thread Size | 13-425 GPI |
| Product Type | Hose Connector |