Learn More
DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks
Tienda Wheaton Science Products Productos
Click to view available options
For Use With (Equipment):
1000mL Celstir Spinner Flask
125mL Celstir Spinner Flask
250mL Celstir Spinner Flask
3000mL Celstir Spinner Flask
500mL Celstir Spinner Flask
50mL Celstir Spinner Flask
Tamaño de la unidad:
Each
Description
- This non-breakable stainless steel shaft, for replacing the glass shaft included with Celstir™ flasks, may be retrofitted to the existing Celstir flasks.
Specifications
Specifications
| For Use With (Equipment) | 1000mL Celstir Spinner Flask |
| Includes | Top cap with liners, glass rod, magnet holder, magnet and stirring paddle |
| Type | Impeller |