Learn More
DWK Life Sciences Wheaton™ LDPE Dispensing Bottles
Product Code. 12485502 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
Unit Size:
Pack of 144
Pack of 72
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12485502 60 mL Pack of 144
12495502 125 mL Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12485502 Supplier DWK Life Sciences Supplier No. 211194
  • ONLINE EXCLUSIVE PRICE
    € 426.80
    € 422.15 / Pack of 144
    Save € 4.65
    1% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
Excellent for mild and strong buffers, good chemical resistance
  • Made from low density polyethylene
  • Translucent color
  • Very flexible with high impact strength
  • Raise the spout to dispense contents and depress spout to seal bottle
TRUSTED_SUSTAINABILITY
Specifications
Capacity 2.03 oz., 60 mL
Capacity (Metric) 60 mL
Capacity (English) 2.03 oz.
Color Natural
Diameter (English) 1.5 in.
Material LDPE
Height (Metric) 92 mm
Product Type Dispensing Bottle
Height (English) 3.6 in.
Closure Size 20-410 mm
Diameter (Metric) 39 mm
Closure Type Screw Cap
Autoclavable Not Autoclavable