Learn More
DWK Life Sciences Wheaton™ LDPE Dropping Bottles With Streaming Tips and Polypropylene Caps
Product Code. 12465512 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
7 mL
15 mL
30 mL
60 mL
Closure Size:
15-415 mm
20-410 mm
Unit Size:
Pack of 144
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size unitSize
12465512 7 mL 15-415 mm Pack of 144
12475512 15 mL 15-415 mm Pack of 144
12485512 30 mL 20-410 mm Pack of 144
12495512 60 mL 20-410 mm Pack of 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12465512 Supplier DWK Life Sciences Supplier No. 211603
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
Tip dispenses a stream of liquid or noncontrolled drop
  • Made of nonautoclavable, low-density polyethylene
  • Dropper bottles, tips and caps are packaged separately
TRUSTED_SUSTAINABILITY
Specifications
Capacity 0.24 oz., 7 mL
Capacity (Metric) 7 mL
Capacity (English) 0.24 oz.
Color Natural
Diameter (English) 0.74 in.
Includes Streaming tip and Polypropylene cap
Material LDPE
Closure Included Yes
Height (Metric) 62 mm
Height (English) 2.44 in.
Closure Material Polypropylene
Closure Size 15-415 mm
Diameter (Metric) 19 mm
Autoclavable Not Autoclavable