Learn More
DWK Life Sciences™ Wheaton™ Polyacetal Clamps
Shop All Wheaton Science Products Products
missing translation for 'orderingAttributeHoverText'
Color:
Blue
Dark Green
Yellow
missing translation for 'unitSize'
Pack of 12
Description
- Molded from polyoxymethylene (POM), an acetal resin
- Will not scratch glass and can withstand temperatures up to 150°C
- Resistant to bases and weak acids
- Not resistant to halogen gases and concentrated acids
Specifications
Specifications
| Type | Standard Taper Clamp |
| Material | POM Acetal |
| For Use With (Equipment) | Kuderna Danish Evaporative Concentrator |
| Color | Blue |