Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Red
Product Code. 12436111 Shop All Wheaton Science Products Products
Click to view available options
For Use With (Equipment):
Dropping bottle
Plastic dropping bottle
Neck Finish:
13-425
15-415
20-410
8-425
No. per Case:
100 Cs.
1000 Cs.
Unit Size:
Pack of 100
Pack of 1000
Product Code. 12436111
Brand: DWK Life Sciences W242512A
Only null left
Add to basket
Add to basket
Screw cap made of radiation grade polypropylene
  • Packaged in case of 100 or 1000
TRUSTED_SUSTAINABILITY
Specifications
Color Red
Material Polypropylene
No. per Case 1000 Cs.
Neck Finish 15-415
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Dropping bottle