Learn More
DWK Life Sciences Wheaton™ Tamper-Evident Caps
Shop All Wheaton Science Products Products
Click to view available options
No. per Case:
2500 Case
5000 Case
Unit Size:
100g
Pack of 5000
Description
- Use with Wheaton Uni-Dose Bottles and Vials
- Three-piece Pulp / PVDC / PE lined aluminum caps have unique metal inserts
- A metal tab with a rolled safety edge “perks up” to provide visual proof of tight seal
- Caps remove quickly and easily in one piece
- Cat. No. 03-084-130 is stamped inside “For Oral Use”
- Attach using Wheaton Crimper
Specifications
Specifications
| Includes | Rolled safety edge |
| Material | Aluminum |
| No. per Case | 5000 Case |
| Liner Material | Pulp/PVDC/Polyethylene |
| Certifications/Compliance | ISO 9001 |
| For Use With (Equipment) | Uni dose bottle and vial |