Learn More
DWK Life Sciences Wheaton™ Tamper-Evident Caps
Product Code. 11714319 Shop All Wheaton Science Products Products
Click to view available options
No. per Case:
2500 Case
5000 Case
Unit Size:
100g
Pack of 5000
Product Code. 11714319
Brand: DWK Life Sciences W224199
Only null left
Add to basket
Add to basket
Contents cannot be accessed without removing cap or leaving evidence of puncture or insert
  • Use with Wheaton Uni-Dose Bottles and Vials
  • Three-piece Pulp / PVDC / PE lined aluminum caps have unique metal inserts
  • A metal tab with a rolled safety edge “perks up” to provide visual proof of tight seal
  • Caps remove quickly and easily in one piece
  • Cat. No. 03-084-130 is stamped inside “For Oral Use”
  • Attach using Wheaton Crimper
TRUSTED_SUSTAINABILITY
Specifications
Includes Rolled safety edge
Material Aluminum
No. per Case 5000 Case
Liner Material Pulp/PVDC/Polyethylene
Certifications/Compliance ISO 9001
For Use With (Equipment) Uni dose bottle and vial