Learn More
AmPRi™ White Med Comfort Latex Powder-Free Non-Sterile Gloves
AmPRi™ Med-Comfort™ Powder-Free Latex Gloves are double chlorinated, resulting in protein values that are almost impossible to detect
Brand:  AmPRi™ 01030 XL
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15251089
  • € 107.61 / Pack of 1000
Estimated Shipment
15-07-2020
Add to basket
Description
Description
  • Exceptionally versatile latex powder-free gloves
  • Double chlorinated, resulting in protein values that are almost impossible to detect
  • Increased thickness for more protection, even with more difficult applications
  • Non-sterile
  • Thickness AQL 1.5
  • Light-abrasive palms
Specifications
Specifications
Yes
Ambidextrous
X-Large
White
AQL 1.5, EN 455, EN420, CE
Beaded
Latex
Powder-free