Learn More
3M™ White Polyethylene Laminate Category I Protective Coveralls
Click to view available options
Size:
2X-Large
3X-Large
4X-Large
Large
Large/X-Large
Medium
Small
X-Large
Unit Size:
Pack of 20
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | 2X-Large |
| Gender | Unisex |
| Disposable | Single-use |
| Waist Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Hood Type | 2-panel |
| Sterility | Non-sterile |
| Protection Class | Category I |
| Show More |