Learn More
- Home
- Products
- Facility Safety and Maintenance
- Labels and Label Makers
- Bottle and Tube Labels
- GA International White Removable Deep-Freeze Labels for Barcode Printers on Small Roll
GA International White Removable Deep-Freeze Labels for Barcode Printers on Small Roll
Product Code. 17879477
Change view
Click to view available options
Dimensions (L x W):
101.6 x 101.6 mm
22.2 x 11 mm
22.2 x 203.2 mm
22.9 x 19 mm
25.4 x 12.7 mm
25.4 x 19.1 mm
25.4 x 25.4 mm
25.4 x 44.5 mm
25.4 x 50.8 mm
25.4 x 7 mm
31.8 x 25.4 mm
32.5 x 15.9 mm
33 x 15.9 mm
38.1 x 63.5 mm
44.5 x 12.7 mm
44.5 x 25.4 mm
50.8 x 25.4 mm
50.8 x 40.6 mm
6.4 x 25.4 mm
63.5 x 25.4 mm
67.1 x 7 mm
76.2 x 25.4 mm
Quantity:
1000/Pk.
10000/Pk.
2000/Pk.
250/Pk.
300/Pk.
3000/Pk.
4000/Pk.
Labels per Row:
1 Label
10 Labels
3 Labels
5 Labels
Unit Size:
Pack of 1000
Pack of 10000
Pack of 2000
Pack of 250
Pack of 300
Pack of 3000
Pack of 4000
25 product options available for selection
| Product Code. | Dimensions (L x W) | Quantity | Labels per Row | unitSize |
|---|---|---|---|---|
| 17879477 | 67.1 x 7 mm | 2000/Pk. | 1 Label | Pack of 2000 |
| 17869677 | 101.6 x 101.6 mm | 300/Pk. | 1 Label | Pack of 300 |
| 17859287 | 67.1 x 7 mm | 4000/Pk. | 1 Label | Pack of 4000 |
| 17891128 | 25.4 x 7 mm | 2000/Pk. | 1 Label | Pack of 2000 |
| 17821328 | 22.9 x 19 mm | 1000/Pk. | 1 Label | Pack of 1000 |
| 17820658 | 22.2 x 11 mm | 2000/Pk. | 1 Label | Pack of 2000 |
| 17880368 | 6.4 x 25.4 mm | 10000/Pk. | 10 Labels | Pack of 10000 |
| 17850558 | 25.4 x 44.5 mm | 1000/Pk. | 1 Label | Pack of 1000 |
| 17848867 | 25.4 x 44.5 mm | 3000/Pk. | 3 Labels | Pack of 3000 |
| 17891188 | 22.2 x 203.2 mm | 250/Pk. | 1 Label | Pack of 250 |
| 17869637 | 33 x 15.9 mm | 1000/Pk. | 1 Label | Pack of 1000 |
| 17810138 | 50.8 x 40.6 mm | 1000/Pk. | 1 Label | Pack of 1000 |
| 17800708 | 6.4 x 25.4 mm | 10000/Pk. | 5 Labels | Pack of 10000 |
| 17859937 | 44.5 x 25.4 mm | 2000/Pk. | 1 Label | Pack of 2000 |
| 17830808 | 50.8 x 25.4 mm | 1000/Pk. | 1 Label | Pack of 1000 |
| 17869827 | 25.4 x 25.4 mm | 2000/Pk. | 1 Label | Pack of 2000 |
| 17880128 | 76.2 x 25.4 mm | 1000/Pk. | 1 Label | Pack of 1000 |
| 17809347 | 44.5 x 12.7 mm | 2000/Pk. | 1 Label | Pack of 2000 |
| 17820808 | 38.1 x 63.5 mm | 1000/Pk. | 1 Label | Pack of 1000 |
| 17841318 | 25.4 x 19.1 mm | 1000/Pk. | 1 Label | Pack of 1000 |
| 17899867 | 31.8 x 25.4 mm | 1000/Pk. | 1 Label | Pack of 1000 |
| 17830358 | 63.5 x 25.4 mm | 1000/Pk. | 1 Label | Pack of 1000 |
| 17880828 | 25.4 x 12.7 mm | 1000/Pk. | 1 Label | Pack of 1000 |
| 17868897 | 25.4 x 50.8 mm | 1000/Pk. | 1 Label | Pack of 1000 |
| 17879757 | 32.5 x 15.9 mm | 1000/Pk. | 1 Label | Pack of 1000 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
Description
- Ideal for long-term storage in deep-freeze conditions (-80°C)
- Removable adhesive ensures the labels can be easily removed without leaving any adhesive residue
- Printable with all major thermal-transfer printer brands
- Resistant to gamma irradiation (tested up to 50 KGy)
Specifications
Specifications
| Adhesive Type | Removable |
| Color | White |
| Finish | Matte |
| Format | Roll |
| Material | Thermoplastic Film |
| Print Technology | Thermal Transfer |
| Product Type | Label |
| Sterility | Non-sterile |
| Labels per Row | 1 Label |
| Quantity | 2000/Pk. |
| Shape | Rectangle |
| Temperature Range (English) | -112°F to +212°F |
| Temperature Range (Metric) | -80°C to +100°C |
| Dimensions (L x W) | 67.1 x 7 mm |
| Length (English) | 0.28 in. |
| Length (Metric) | 7 mm |
| Width (English) | 2.64 in. |
| Width (Metric) | 67.1 mm |
| Diameter (English) | 1 in. |
| Diameter (Metric) | 25.4 mm |
| Show More Show Less |