Learn More
Atrow™ White Stainless Steel Tube Rack
Click to view available options
Positions:
24
Unit Size:
Each
Specifications
Specifications
| Color | White |
| Holds | Tubes (16 to 20 mm) |
| Material | Stainless Steel |
| Positions | 24 |
| For Use With (Equipment) | For Hemolysis Tube |
| No. of Racks | 4 x 6 |
| No. per Case | 24 |
| Diameter (Metric) Opening | 20 mm |
| Product Type | Test Tube Rack |
| Height (Metric) | 65 mm |
| Show More |