Learn More
Kimberly-Clark™ White Wypall X70 Cloth
Ideal for medium-duty maintenance. Kimberly Clark™ White Wypall X70 Cloth Available as quarter folded wipers wrapped in a splash-proof, water-repellent polybag.
Brand:  Kimberly-Clark™ 8384
Additional Details : Weight : 5.19144kg
 View more versions of this product
Product Code. 10014660
  • € 125.46 / 1 roll
In stock, ready to ship: 8
Add to basket
Description
Description
  • A repeat-use wiper with HYDROKNIT technology
  • Highly absorbent, strong and durable (wet or dry)
  • Resists most solvents, reduces residue, allows fast cleaning and helps reduce costs
Specifications
Specifications
420mm
380 x 420mm
380 mm
Wypall X70 Cloth
1 Roll x 500 Sheets
White
For medium-duty maintenance, wiping away oil, grease, grime, solvents
Hydroknit™ Fabric
Wypall wiper dispenser
1-Ply