missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WHSC1L1 Monoclonal Antibody (OTI1F11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809975
This item is not returnable.
View return policy
Description
This gene is related to the Wolf-Hirschhorn syndrome candidate-1 gene and encodes a protein with PWWP (proline-tryptophan-tryptophan-proline) domains. The function of the protein has not been determined. Two alternatively spliced variants have been described.Specifications
| WHSC1L1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BZ95 | |
| NSD3 | |
| Human recombit protein fragment corresponding to amino acids 427-566 of human WHSC1L1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1F11 | |
| Unconjugated | |
| NSD3 | |
| KMT3F, KMT3G, WHISTLE, WHSC1L1, pp14328 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54904 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction