missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WHSC1L1 Monoclonal Antibody (OTI1F11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809975S
This item is not returnable.
View return policy
Description
WHSC1L1 Monoclonal antibody specifically detects WHSC1L1 in Human samples. It is validated for Western BlotSpecifications
| WHSC1L1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P78545 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1999 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1F11 | |
| Unconjugated | |
| ELF3 | |
| 6720429E03; A530023P05; AI528490; DC28; DKFZp667H044; FLJ20353; Histone-lysine N-methyltransferase NSD3; MGC126766; MGC142029; NSD3; nuclear receptor binding SET domain protein 3; nuclear SET domain-containing protein 3; pp14328; Protein whistle; WHISTLE; WHSC1L1; WHSC1-like 1 isoform 9 with methyltransferase activity to lysine; WHSC1-like protein 1; Wolf-Hirschhorn syndrome candidate 1-like 1; Wolf-Hirschhorn syndrome candidate 1-like 1 (human); Wolf-Hirschhorn syndrome candidate 1-like protein 1; Wolf-Hirschhorn syndrome candidate 1-like protein 1 homolog | |
| Human recombinant protein fragment corresponding to amino acids 175-286 of human ELF3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction