missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIBG Antibody (OTI5E2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74868B
This item is not returnable.
View return policy
Description
WIBG Monoclonal specifically detects WIBG in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| WIBG | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| PYM1 | |
| Full length recombinant protein of human WIBG (NP_115721) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5E2 | |
| Western Blot, Immunohistochemistry | |
| MGC13064, partner of Y14 and mago, PYMProtein wibg homolog, within bgcn homolog (Drosophila) | |
| Mouse | |
| Affinity Purified | |
| DNA replication Transcription Translation and Splicing | |
| 84305.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction