missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIBG Monoclonal Antibody (OTI5E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806496S
This item is not returnable.
View return policy
Description
WIBG Monoclonal antibody specifically detects WIBG in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| WIBG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P10276 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5914 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5E2 | |
| Unconjugated | |
| RARA | |
| A030010B05Rik; Partner of Y14 and mago; protein wibg homolog; PYM; PYM homolog 1 exon junction complex-associated factor; PYM homolog 1, exon junction complex associated factor; Pym1; RGD1306096; WIBG; within bgcn homolog; within bgcn homolog (Drosophila) | |
| Human recombinant protein fragment corresponding to amino acids 52-299 of human RARA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction