Learn More
Cole-Parmer™ Wide-Mouth Soda Lime Glass Sample Jars
Catalog Number p-8604019
Click to view available options
Capacity:
500 mL
960 mL
Diameter (Metric):
9.1 cm
9.5 cm
Height (Metric):
9.6 cm
17 cm
Precleaned wide-mouth glass jars well suited for semivolatiles, pesticides, PCBs, and metals. Cole-Parmer™ Wide-Mouth Soda Lime Glass Sample Jars are ideal for environmental sampling.
  • Manufactured from USP Type III soda-lime glass
  • Includes chemically resistant PTFE liner inserted and screw cap
  • Sample jars include a certificate of analysis.
Capacity 500 mL
Closure Liner PTFE
Closure Type Screw Cap
Diameter (Metric) 9.1 cm
Height (English) 3.75 in.
Height (Metric) 9.6 cm
Product Type Sample Jar
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Clear
Diameter (English) 3.5 in.
For Use With (Application) Semivolatiles, Pesticides, PCBS, and Metals, Ideal for Environmental Sampling
Includes Screw Cap
Material Glass
No. per Pack 12/Pk
Weight (Metric) 0.8 kg
Clarity Clear
Certifications/Compliance Manufactured from USP Type III Soda Lime Glass
Autoclavable Autoclavable
Mouth Wide
Shape Round