Learn More
Duran™ Wide Neck Soda Lime Glass Reagent Bottless with Stopper
Manufactured from soda lime glass. DURAN™ Wide Neck Soda Lime Glass Reagent Bottless with Stopper is DIN 12039 certified.
Merk: Duran™ 231853606
Overige gegevens : Gewicht : 2.50000kg
Beschrijving
- Wide neck
- Clear
- With individually ground glass stopper
- Soda-lime-glass
- DIN 12039
Specificatie
250 ml | |
10 per pak | |
Reagentiavat | |
129 mm |
Soda Lime Glass | |
NS 34/24 | |
71 mm |