missing translation for 'onlineSavingsMsg'
Learn More

WIF-1 Antibody (JE41-39), Novus Biologicals™

Product Code. 30116760 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30116760 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30116760 Supplier Novus Biologicals Supplier No. NBP333102

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

WIF-1 Monoclonal antibody specifically detects WIF-1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen WIF-1
Applications Western Blot
Classification Monoclonal
Clone JE41-39
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol
Gene Alias WIF-1, WNT inhibitory factor 1
Host Species Rabbit
Immunogen Synthetic peptide within Human WIF-1 aa 330-379/ 379. (Uniprot: Q9Y5W5)
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 11197
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.