Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Shigemi™ Susceptibility Matched NMR Tubes
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Shigemi™ Susceptibility Matched NMR Tubes feature a susceptibility matched bottom and plunger to effectively reduce the boundary gradients at the edges of the Rf coil reducing the required sample volume by two thirds.
€ 305.00 - € 357.00
Product Code | Brand | Diameter (Metric) Outer | For Use With (Equipment) | Model | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | For Use With (Equipment) | Model | Price | Quantity & Availability | |||
15388588
|
Bel-Art™ SP Scienceware™
66000-0027 |
3 mm | Universal, All Spectometers | BMS-003 |
€ 309.00
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
15398588
|
Bel-Art™ SP Scienceware™
66000-0028 |
5 mm | Bruker Spectometers | BMS-005B |
€ 305.00
Each |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||
15338598
|
Bel-Art™ SP Scienceware™
66000-0030 |
3 mm | Universal, All Spectometers | CMS-003 |
€ 312.00
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
15358598
|
Bel-Art™ SP Scienceware™
66000-0031 |
5 mm | Bruker Spectometers | CMS-005B |
€ 309.00
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
15368598
|
Bel-Art™ SP Scienceware™
66000-0032 |
3 mm | Universal, All Spectometers | DMS-003 |
€ 357.00
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
15308608
|
Bel-Art™ SP Scienceware™
66000-0036 |
3 mm | Universal, All Spectometers | MMS-003 |
€ 357.00
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
Description
- Select tube based on solvent used: CDCL3, CD3OD, D2O, DMSO-d6
- Available for Bruker or Universal
Specifications
Tube Set |