Learn More
Gibertini Elettronica™ Super Dee Digital Distilling Unit
Description
Control panel with Alphanumeric Intelligent Display and keyboard drives and controls the microprocessor for:
- The regulation and the tare of the new balance
- The volume to be distilled
- The real time value of the distilled amount
- Check of the inlet cooling water: temperature, pressure and flow.
- Functional requirement: T max = 20°C – press min = 1,5 bar – flow min = 5L/min
- The different languages (IT/ES/UK/FR)
- The warning of missing glassware
- The warning of opened protection or drainage
Integrated Steam Generator: no more distilled water required. Direct connection to the tap water and cartridge for demineralisation (endurance 100L – ca 400 volatile acidity analyses) interchangeable by the user. Available also without steam generator.
- AISI 316 stainless steel framework
- New vertical sliding Plexiglas protection
- New phial with enlarged drainage to distillate wine lees and grape pulp
- New system rapid and operational (one hand operation) for holding the glassware. It inhibits the distillation if the glassware is not present
- New shower for washing the phial with flow regulation
- Magnetic filter at the inlet of the water for stopping heavy particles
- Automatic stop of the instrument after the distillation helps conserve water
Specifications
Specifications
| Electrical Requirements | 230V, 50/60Hz |
| Voltage | 230V, 50/60Hz |
| Includes | 3 bottles 100mL, 200mL vial 1, 2 300mL conical flasks, 1 wash bottle 500mL, 3 wash bottles, chemical reagents and demineralization cartridge |
| Depth (Metric) | 300mm |
| Height (Metric) | 1200mm |
| Type | Wine Distiller |
| Material | Stainless steel |
| For Use With (Equipment) | wines, spirits, beers, perfumes and alcoholic solutions |
| Quantity | 1Each |
| Dimensions (L x W x H) | 300 x 290 x 1200mm |
| Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.