Learn More
Bode Chemie™ Wipes
Product Code. 10433475
Change view
Click to view available options
Dimensions (L x W):
180 x 200 mm
Unit Size:
80 sheets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10433475 180 x 200 mm 80 sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10433475 Supplier Bode Chemie™ Supplier No. 9813875
  • ONLINE EXCLUSIVE PRICE
    € 103.00
    € 8.68 / 80 sheets
    Save € 94.32
    91% Off
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Wipes
Dimensions (L x W) 180 x 200 mm
For Use With (Application) Cleaning disinfection of alcohol-sensitive surfaces
For Use With (Equipment) sensitive medical devices