missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIPI1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25204-1-AP-150UL
This item is not returnable.
View return policy
Description
WIPI1 (WD repeat domain, phosphoinositide interacting-1), also known as WIPI1, ATG18 or WIPI49, is thought to play a role in autophagy and may regulate protein trafficking in certain recycling pathways. It contains three WD repeats and has a 7-bladed propeller structure with a conserved motif that facilitates its interaction with other proteins. WIPI1 localizes to cytoplasmic vesicles, endosomes, clathrin-coated vesicles and the trans-Golgi network. It is ubiquitously expressed with highest expression in heart, testis, placenta, pancreas and skeletal muscle. WIPI1 is upregulated in a variety of tumors, suggesting a role in carcinogenesis.Specifications
| WIPI1 | |
| Polyclonal | |
| Unconjugated | |
| WIPI1 | |
| ATG18, Atg18 protein homolog, WIPI 1, WIPI 49 kDa, WIPI1, WIPI49 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 52639, 55062 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5MNZ9, Q8R3E3 | |
| Wipi1 | |
| WIPI1 Fusion Protein Ag18461 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction